Learn More
Medchemexpress LLC Diaveridine | 5355-16-8 | MFCD00057349 | 99.8% | 5 G
SDP
Catalog No. 5000412207
Encompass_Preferred
Diaveridine is a dihydrofolate reductase (DHFR) inhibitor with a Ki of 11.5 nM for the wild type DHFR and also an antibacterial agent. It is intended for research use only.
  • Dihydrofolate reductase (DHFR) inhibitor with a Ki of 11.5 nM
  • Functions as an antibacterial agent
  • Exhibits strong bactericidal effect on S. typhimurium TA1535 at 10 μg/mL or more
  • Causes a dose-dependent, significant increase in aberrant metaphases at 100 μg/mL
  • Does not cause abnormal chromosome aberration at studied doses
  • Does not produce significant changes in organ-to-body weight ratio
Catalog No. 50-004-12207 Supplier Medchemexpress LLC Supplier No. HYB19025G
Diaveridine is a dihydrofolate reductase (DHFR) inhibitor with a Ki of 11.5 nM for the wild type DHFR and also an antibacterial agent. It is intended for research use only.
  • Dihydrofolate reductase (DHFR) inhibitor with a Ki of 11.5 nM
  • Functions as an antibacterial agent
  • Exhibits strong bactericidal effect on S. typhimurium TA1535 at 10 μg/mL or more
  • Causes a dose-dependent, significant increase in aberrant metaphases at 100 μg/mL
  • Does not cause abnormal chromosome aberration at studied doses
  • Does not produce significant changes in organ-to-body weight ratio