Learn More
Medchemexpress LLC Diaza-15-crown-5 100mg
SDP
Catalog No. 5000206958
Encompass_Preferred
Diaza-15-crown-5 extends the lifetime of M intermediates in BR membranes Diaza-15-crown-5 is a crown ether compound that alters the surface charge of BR membranes Diaza-15-crown-5 changes the surface charge of the BR film and overall membrane stability Diaza-15-crown-5 allows the M state to be maintained for a longer period of time[1]
Catalog No. 50-002-06958 Supplier Medchemexpress LLC Supplier No. HYW102545100MG
Diaza-15-crown-5 extends the lifetime of M intermediates in BR membranes Diaza-15-crown-5 is a crown ether compound that alters the surface charge of BR membranes Diaza-15-crown-5 changes the surface charge of the BR film and overall membrane stability Diaza-15-crown-5 allows the M state to be maintained for a longer period of time[1]