Learn More
AdipoGen Diazidostilbenedisulfonic acid
SDP
Catalog No. 502561313
Encompass_Preferred
Chemical. CAS 2718-90-3. Formula C14H8N6Na2O6S2 . 4H2O. MW 538.42. Synthetic. A reagent for enzyme immobilisation on fibroin by photo-crosslinking. The reagent is water-soluble over a wide pH range.
Catalog No. 50-256-1313 Supplier AdipoGen Supplier No. CDXD0022G010
Chemical. CAS 2718-90-3. Formula C14H8N6Na2O6S2 . 4H2O. MW 538.42. Synthetic. A reagent for enzyme immobilisation on fibroin by photo-crosslinking. The reagent is water-soluble over a wide pH range.