Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diazine Black, MP Biomedicals

Catalog No. ICN213956 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN213956 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN213956 Supplier MP Biomedicals, Inc Supplier No. 0521395610

Chemical Identifiers

CAS 4443-99-6
Molecular Formula C28H26ClN5O
Molecular Weight (g/mol) 484
InChI Key VJDDAARZIFHSQY-UHFFFAOYSA-N
PubChem CID 9891477
IUPAC Name 4-[[8-(diethylamino)-10-phenylphenazin-10-ium-2-yl]hydrazinylidene]cyclohexa-2,5-dien-1-one;chloride
SMILES CCN(CC)C1=CC2=[N+](C3=C(C=CC(=C3)NN=C4C=CC(=O)C=C4)N=C2C=C1)C5=CC=CC=C5.[Cl-]

Specifications

Quantity 10 g
Chemical Name or Material Diazine Black
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.