Learn More
Medchemexpress LLC Diazinon | 333-41-5 | 99.25% | 250 MG
SDP
Catalog No. 5000413948
Encompass_Preferred
Diazinon is an orally active, irreversible acetylcholinesterase (AChE) inhibitor and insecticide. It is primarily used in agriculture and can be absorbed through the digestive system, skin, or respiratory tract. It affects the nervous system by inhibiting AChE, leading to acetylcholine accumulation and overstimulation of ACh receptors. Diazinon also produces reactive oxygen species, inducing oxidative stress in various tissues, and has potential effects on human and animal health.
  • Inhibits acetylcholinesterase (AChE)
  • Induces oxidative stress through reactive oxygen species (ROS) production
  • Suitable for agricultural use as an insecticide
  • Inhibits cell viability and induces cell cycle arrest in specific cell lines *in vitro*
  • Causes renal damage and suppresses immune activities in animal models *in vivo*
Catalog No. 50-004-13948 Supplier Medchemexpress LLC Supplier No. 250MGHYB1113
Add to Cart
Add to Cart
Diazinon is an orally active, irreversible acetylcholinesterase (AChE) inhibitor and insecticide. It is primarily used in agriculture and can be absorbed through the digestive system, skin, or respiratory tract. It affects the nervous system by inhibiting AChE, leading to acetylcholine accumulation and overstimulation of ACh receptors. Diazinon also produces reactive oxygen species, inducing oxidative stress in various tissues, and has potential effects on human and animal health.
  • Inhibits acetylcholinesterase (AChE)
  • Induces oxidative stress through reactive oxygen species (ROS) production
  • Suitable for agricultural use as an insecticide
  • Inhibits cell viability and induces cell cycle arrest in specific cell lines *in vitro*
  • Causes renal damage and suppresses immune activities in animal models *in vivo*