Learn More
Medchemexpress LLC Diazinon (Dimpylate) | 333-41-5 | 99.3% | 500 MG
SDP
Catalog No. 5000413718
Encompass_Preferred
Diazinon is an orally active, irreversible AChE inhibitor and insecticide that can be absorbed through the digestive system, skin, or respiratory tract. It inhibits AChE, leading to the accumulation of acetylcholine, which overstimulates ACh receptors and affects the nervous system. Diazinon also produces reactive oxygen species (ROS), inducing oxidative stress in various tissues. It is primarily used in agriculture as an insecticide and may have potential effects on human and animal health.
  • Orally active, irreversible AChE inhibitor and insecticide
  • Absorbed through the digestive system, skin, or respiratory tract
  • Inhibits AChE, causing acetylcholine accumulation and overstimulation of ACh receptors
  • Affects the nervous system
  • Produces reactive oxygen species (ROS), inducing oxidative stress
  • Mainly used in the agricultural field as an insecticide
Catalog No. 50-004-13718 Supplier Medchemexpress LLC Supplier No. HYB1113500MG
Diazinon is an orally active, irreversible AChE inhibitor and insecticide that can be absorbed through the digestive system, skin, or respiratory tract. It inhibits AChE, leading to the accumulation of acetylcholine, which overstimulates ACh receptors and affects the nervous system. Diazinon also produces reactive oxygen species (ROS), inducing oxidative stress in various tissues. It is primarily used in agriculture as an insecticide and may have potential effects on human and animal health.
  • Orally active, irreversible AChE inhibitor and insecticide
  • Absorbed through the digestive system, skin, or respiratory tract
  • Inhibits AChE, causing acetylcholine accumulation and overstimulation of ACh receptors
  • Affects the nervous system
  • Produces reactive oxygen species (ROS), inducing oxidative stress
  • Mainly used in the agricultural field as an insecticide