Learn More
TARGETMOL CHEMICALS INC DIAZIQUONE 5MG
SDP
Catalog No. 502579523
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Diaziquone (NSC-182986) is a water-soluble synthetic azacyclic benzoquinone that enhances penetration of the blood-brain barrier has antitumor activity and is used in the treatment of extreme renal failure. purity: 98%
Catalog No. 50-257-9523 Supplier TARGETMOL CHEMICALS INC Supplier No. T613835MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Diaziquone (NSC-182986) is a water-soluble synthetic azacyclic benzoquinone that enhances penetration of the blood-brain barrier has antitumor activity and is used in the treatment of extreme renal failure. purity: 98%