Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DiAzKs (H-L-photo-lysine) | 1253643-88-7 | 99.9% | 272.30 g/mol | C11H20N4O4 | 10 MG
SDP

Catalog No. 5000297703
Encompass_Preferred

DiAzKs (H-L-photo-lysine) is a diazirine-containing lysine derivative used as a UV-activated photo-crosslinking probe for capturing and mapping protein-protein interactions both in vitro and in living cells.

  • Photo-activatable amino acid enabling covalent capture of interacting proteins.
  • Site-selective incorporation into proteins for targeted crosslinking experiments.
  • High purity at 99.9% for reliable experimental results.
  • Molecular weight 272.30 g/mol and chemical formula C11H20N4O4.
  • CAS number 1253643-88-7 for unambiguous identification.
  • Store protected from light at 4°C; stable longer-term in solution at -80°C.

Catalog No. 50-002-97703 Supplier Medchemexpress LLC Supplier No. HYD085310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DiAzKs (H-L-photo-lysine) is a diazirine-containing lysine derivative used as a UV-activated photo-crosslinking probe for capturing and mapping protein-protein interactions both in vitro and in living cells.

  • Photo-activatable amino acid enabling covalent capture of interacting proteins.
  • Site-selective incorporation into proteins for targeted crosslinking experiments.
  • High purity at 99.9% for reliable experimental results.
  • Molecular weight 272.30 g/mol and chemical formula C11H20N4O4.
  • CAS number 1253643-88-7 for unambiguous identification.
  • Store protected from light at 4°C; stable longer-term in solution at -80°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.