Learn More
Medchemexpress LLC DiAzKs hydrochlorid 10mM 1mL | 2421187-79-1 | 98.0% | 10MM/1ML
SDP
Catalog No. 5000409840
Encompass_Preferred
DiAzKs (H-L-Photo-lysine) hydrochloride is a diazirine-containing lysine amino acid and a photo-cross-linker. It can be site-selectively incorporated into proteins and is used to crosslink protein-protein interactions in vitro and in living cells. This compound acts as a UV light-activated photo-crosslinking probe.
  • Site-selective incorporation into proteins.
  • Crosslinks protein-protein interactions.
  • UV light-activated photo-crosslinking probe.
  • Used in vitro and in living cells.
Catalog No. 50-004-09840 Supplier Medchemexpress LLC Supplier No. HYD0853A10MM/1ML
DiAzKs (H-L-Photo-lysine) hydrochloride is a diazirine-containing lysine amino acid and a photo-cross-linker. It can be site-selectively incorporated into proteins and is used to crosslink protein-protein interactions in vitro and in living cells. This compound acts as a UV light-activated photo-crosslinking probe.
  • Site-selective incorporation into proteins.
  • Crosslinks protein-protein interactions.
  • UV light-activated photo-crosslinking probe.
  • Used in vitro and in living cells.