Learn More
Medchemexpress LLC Diazo biotin-PEG3-azide | 1339202-33-3 | 95.5% | C33H45N9O7S | 25 MG
SDP
Catalog No. 5000352856
Encompass_Preferred
Diazo Biotin-PEG3-azide is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring an Azide group. This allows it to participate in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules containing Alkyne groups, and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules that have DBCO or BCN groups. PROTACs themselves consist of two different ligands linked together, one targeting an E3 ubiquitin ligase and the other a specific protein, leveraging the intracellular ubiquitin-proteasome system to degrade target proteins selectively.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Features an azide group
  • Participates in copper-catalyzed azide-alkyne cycloaddition reactions
  • Undergoes strain-promoted alkyne-azide cycloaddition reactions
Catalog No. 50-003-52856 Supplier Medchemexpress LLC Supplier No. 25MGHY140918
Diazo Biotin-PEG3-azide is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring an Azide group. This allows it to participate in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules containing Alkyne groups, and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules that have DBCO or BCN groups. PROTACs themselves consist of two different ligands linked together, one targeting an E3 ubiquitin ligase and the other a specific protein, leveraging the intracellular ubiquitin-proteasome system to degrade target proteins selectively.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Features an azide group
  • Participates in copper-catalyzed azide-alkyne cycloaddition reactions
  • Undergoes strain-promoted alkyne-azide cycloaddition reactions