Learn More
Medchemexpress LLC Diazo biotin-PEG3-azide | 1339202-33-3 | MFCD28334530 | 95.5% | C33H45N9O7S | 10MG
SDP
Catalog No. 5000259695
Encompass_Preferred
Diazo biotin-PEG3-azide is a multifunctional PEG3 linker combining a cleavable diazo functional group, a biotin affinity tag, and an azide reactive handle. It is designed for efficient biotinylation and capture of alkyne-bearing biomolecules using click chemistry and for use as a linker in chemical biology workflows.
  • Provides azide functionality for CuAAC and SPAAC click reactions.
  • Includes a biotin tag for affinity purification and detection.
  • Contains a diazo group enabling photo-cleavage or crosslinking.
  • PEG3 spacer improves solubility and reduces steric hindrance.
  • Suitable for biotinylation, pull-down assays, and linker synthesis.
Catalog No. 50-002-59695 Supplier Medchemexpress LLC Supplier No. HY14091810MG
Diazo biotin-PEG3-azide is a multifunctional PEG3 linker combining a cleavable diazo functional group, a biotin affinity tag, and an azide reactive handle. It is designed for efficient biotinylation and capture of alkyne-bearing biomolecules using click chemistry and for use as a linker in chemical biology workflows.
  • Provides azide functionality for CuAAC and SPAAC click reactions.
  • Includes a biotin tag for affinity purification and detection.
  • Contains a diazo group enabling photo-cleavage or crosslinking.
  • PEG3 spacer improves solubility and reduces steric hindrance.
  • Suitable for biotinylation, pull-down assays, and linker synthesis.