Learn More
Selleck Chemical LLC Diazolidinyl urea S5292-25mg
SDP
Catalog No. 503112214
Encompass_Preferred
Diazolidinyl urea (Germall II) is an antimicrobial preservative used in cosmetics
Catalog No. 50-311-2214 Supplier Selleck Chemical LLC Supplier No. S529225MG
Diazolidinyl urea (Germall II) is an antimicrobial preservative used in cosmetics