Learn More
Medchemexpress LLC HY-W009350 500mg Medchemexpress, Diazolidinyl urea CAS:78491-02-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW009350500MG
Encompass_Preferred
Medchemexpress, HY-W009350 500mg Diazolidinyl urea CAS:78491-02-8 Diazolidinyl urea, a broad spectrum preservative, is a formaldehyde-releasing compound that releases formaldehyde through its decomposition. Diazolidinyl urea is effective against most contaminating microorganisms, especially Pseudomonas. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3270