Learn More
Medchemexpress LLC Diazolidinyl urea | 78491-02-8 | 95.0% | 278.22 | 1 G
SDP
Catalog No. 5000453086
Encompass_Preferred
Diazolidinyl urea is a broad-spectrum preservative commonly used in cosmetics. It exerts antibacterial effects by decomposing and releasing formaldehyde. It is effective against most contaminating microorganisms, especially Pseudomonas.
  • Broad-spectrum preservative
  • Commonly used in cosmetics
  • Exerts antibacterial effects by decomposing and releasing formaldehyde
  • Effective against most contaminating microorganisms, especially *Pseudomonas*
  • Targets bacterial pathogens
  • Anti-infection pathway
Catalog No. 50-004-53086 Supplier Medchemexpress LLC Supplier No. HYW0093501G
Diazolidinyl urea is a broad-spectrum preservative commonly used in cosmetics. It exerts antibacterial effects by decomposing and releasing formaldehyde. It is effective against most contaminating microorganisms, especially Pseudomonas.
  • Broad-spectrum preservative
  • Commonly used in cosmetics
  • Exerts antibacterial effects by decomposing and releasing formaldehyde
  • Effective against most contaminating microorganisms, especially *Pseudomonas*
  • Targets bacterial pathogens
  • Anti-infection pathway