Learn More
Medchemexpress LLC Diazolidinyl urea | 78491-02-8 | 95.0% | 278.22 | 1 ML
SDP
Catalog No. 5000366854
Encompass_Preferred
Diazolidinyl urea is a broad-spectrum preservative commonly used in cosmetics. It functions by decomposing and releasing formaldehyde, which provides antibacterial effects. It is effective against most contaminating microorganisms, particularly Pseudomonas. This product is intended for research use only.
  • Broad-spectrum preservative for cosmetics
  • Provides antibacterial effects through formaldehyde release
  • Effective against most contaminating microorganisms, including Pseudomonas
  • Shows antimicrobial activity against P. aeruginosa, S. marcescens, and S. epidermidis in vitro
Catalog No. 50-003-66854 Supplier Medchemexpress LLC Supplier No. HYW00935010MM/1ML
Diazolidinyl urea is a broad-spectrum preservative commonly used in cosmetics. It functions by decomposing and releasing formaldehyde, which provides antibacterial effects. It is effective against most contaminating microorganisms, particularly Pseudomonas. This product is intended for research use only.
  • Broad-spectrum preservative for cosmetics
  • Provides antibacterial effects through formaldehyde release
  • Effective against most contaminating microorganisms, including Pseudomonas
  • Shows antimicrobial activity against P. aeruginosa, S. marcescens, and S. epidermidis in vitro