Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diazolidinyl urea, MP Biomedicals™

Catalog No. ICN19019180 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19019180 100 g
ICN19019125 25 g
ICN19019183 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19019180 Supplier MP Biomedicals, Inc Supplier No. 0219019180
Add to Cart
Add to Cart

Chemical Identifiers

CAS 78491-02-8
Molecular Formula C8H14N4O7
Molecular Weight (g/mol) 278.221
InChI Key SOROIESOUPGGFO-UHFFFAOYSA-N
Synonym diazolidinylurea, diazolidinyl urea, 1-1,3-bis hydroxymethyl-2,5-dioxoimidazolidin-4-yl-1,3-bis hydroxymethyl urea, germall ii, germall 11, imidazolidinyl urea 11, n,n'-bis hydroxymethyl urea, dsstox_cid_9559, dsstox_rid_78776
PubChem CID 62277
IUPAC Name 1-[1,3-bis(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl]-1,3-bis(hydroxymethyl)urea
SMILES C(NC(=O)N(CO)C1C(=O)N(C(=O)N1CO)CO)O

Specifications

Color White
Quantity 100 g
Formula Weight 278.2
Physical Form Powder
Chemical Name or Material Diazolidinyl urea
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.