Learn More
Enzo Life Sciences Diazoxide (1g). CAS: 364-98-7
SDP
Catalog No. 502009511
Encompass_Preferred
ATP-sensitive K+ channel activator. Effective concentrations in patch clamp experiments: 100-600µM. Blocks desensitization of AMPA receptors. Acts in pancreatic β-cells, where it is more potent than pinacidil (ALX-550-285). Antihypertensive and vasodilatory agent. Alternative name: 7-Chloro-3-methyl-2H-1,2,4-benzothiadiazine 1,1-dioxide. Purity: ≥98% (HPLC). Appearance: White to off-white powder. Solubility: Soluble in DMSO (25mg/ml); insoluble in water. Long Term Storage: +4°C.
Catalog No. 50-200-9511 Supplier Enzo Life Sciences Supplier No. ALX550260G001
Add to Cart
Add to Cart
ATP-sensitive K+ channel activator. Effective concentrations in patch clamp experiments: 100-600µM. Blocks desensitization of AMPA receptors. Acts in pancreatic β-cells, where it is more potent than pinacidil (ALX-550-285). Antihypertensive and vasodilatory agent. Alternative name: 7-Chloro-3-methyl-2H-1,2,4-benzothiadiazine 1,1-dioxide. Purity: ≥98% (HPLC). Appearance: White to off-white powder. Solubility: Soluble in DMSO (25mg/ml); insoluble in water. Long Term Storage: +4°C.