Learn More
Enzo Life Sciences Diazoxide (500mg). CAS: 364-98-7
SDP
Catalog No. 502009512
Encompass_Preferred
ATP-sensitive K+ channel activator. Effective concentrations in patch clamp experiments: 100-600µM. Blocks desensitization of AMPA receptors. Acts in pancreatic β-cells, where it is more potent than pinacidil (ALX-550-285). Antihypertensive and vasodilatory agent. Alternative name: 7-Chloro-3-methyl-2H-1,2,4-benzothiadiazine 1,1-dioxide. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (25mg/ml); insoluble in water. Long Term Storage: +4°C.
Catalog No. 50-200-9512 Supplier Enzo Life Sciences Supplier No. ALX550260M500
ATP-sensitive K+ channel activator. Effective concentrations in patch clamp experiments: 100-600µM. Blocks desensitization of AMPA receptors. Acts in pancreatic β-cells, where it is more potent than pinacidil (ALX-550-285). Antihypertensive and vasodilatory agent. Alternative name: 7-Chloro-3-methyl-2H-1,2,4-benzothiadiazine 1,1-dioxide. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (25mg/ml); insoluble in water. Long Term Storage: +4°C.