Learn More
Cayman Chemical Diazoxide-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 1432063-51-8, 1 mg
SDP
Catalog No. 502989286
Encompass_Preferred
An internal standard for the quantification of diazoxide by GC- or LC-MS
Catalog No. 50-298-9286 Supplier Cayman Chemical Supplier No. 299671 MG
An internal standard for the quantification of diazoxide by GC- or LC-MS