Learn More
Abcam Diazo x ide, KIR6. x (ATP-sensitive K+ channel) activator, 100MG
Catalog No. 502707735
Encompass_Preferred
MW 230.67 Da, Purity >99%. Selective KIR6.x (KATP) activator; antihypertensive. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7735 Supplier Abcam Supplier No. AB120266100MG
MW 230.67 Da, Purity >99%. Selective KIR6.x (KATP) activator; antihypertensive. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement