Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beckman Coulter Diazyme Procalcitonin Assay

Catalog No. 21280082 Shop All Beckman Coulter Products
Change view
Click to view available options
For Use With (Equipment):
Beckman Coulter Analyzer
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
21-280-082 Beckman Coulter Analyzer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-280-082 Supplier Beckman Coulter Supplier No. DZ558AKY1
Add to Cart
Add to Cart
This item is not returnable. View return policy

Procalcitonin assay offers exceptional clinical quality for sepsis testing

  • In conjunction with other laboratory findings and clinical assessments, aids in the risk assessment of critically ill patients on their first day of Intensive Care Unit (ICU) admission for progression to severe sepsis and septic shock.
  • High Concordance to established methods at key cut-offs
  • State of the art sensitivity and low-end precision
  • Rapid access to results
  • Minimal sample volume
TRUSTED_SUSTAINABILITY

Specifications

Detectable Analytes Procalcitonin
Type Reagent
For Use With (Equipment) Beckman Coulter Analyzer
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.