Learn More
Biotium DiB
Catalog No. 501964729
Encompass_Preferred
DiB is a lipophilic cationically charged blue fluorescent dye useful for staining cytoplasmic membranes. Properties: Ex/Em (MeOH) = 353/442 nm; MW: 1074 DiB is also available as a ready-to-use solution as CellBrite™ Blue Cytoplasmic Membrane Dye.
Catalog No. 50-196-4729 Supplier Biotium Supplier No. 60036
DiB is a lipophilic cationically charged blue fluorescent dye useful for staining cytoplasmic membranes. Properties: Ex/Em (MeOH) = 353/442 nm; MW: 1074 DiB is also available as a ready-to-use solution as CellBrite™ Blue Cytoplasmic Membrane Dye.