Learn More
AdipoGen DIB-Cl
SDP
Supplier:  AdipoGen CDXD0009M200
Encompass_Preferred
Chemical. CAS 162756-62-9. Formula C22H15ClN2O. MW 358.82. Synthetic. 4-4,5-diphenyl-1H-imidazol-2-ylbenzoyl chloride DIB-Cl as reagent for amine labelling was used successfully for derivatization and fluorescence detection of enantiomers of methamphetamin and p-hydroxymethamphetamin, in urine samples. It has also been used for 3,4-Methylendioxy-N-methylamphetamin in blood, morpholine and orther compounds. DIB-Cl as a label was used on fluorescent derivatization some hydroxyl group containing endocrine disruptors, including bisphenol A BPA, bisphenol B BPB, bisphenol E BPE, bisphenol F BPF and 4-nonylphenol 4-NP. DIB-Cl showed superior fluorescence intensity FI to other commercially available reagents, excellent stability in solution and high reactivity with primary and secondary amines under mild conditions.
Catalog No. 50-256-1324