Learn More
Cole-Parmer DIBA OMNIFIT BOTTLE THRD ADPTR
Catalog No. NC2226368 Shop All Cole-Parmer Products
Encompass_Preferred
Diba Omnifit Bottle Thread Adapter, S40 (Female) to GL45 (Male), PTFE
This item is not returnable. View return policy
Catalog No. NC2226368 Supplier Cole-Parmer Supplier No. 2194299
Diba Omnifit Bottle Thread Adapter, S40 (Female) to GL45 (Male), PTFE