Learn More
Biotium DIBAC4(3)
Catalog No. 501964732
Encompass_Preferred
DiBAC4(3) (Bis-(1,3-Dibarbituric acid)-trimethine oxanol) is a so-called translational membrane potential dye that redistributes within the cell membrane when membrane potential changes. The fluorescence of the dye is enhanced when the dye enters the cell membrane as a result of membrane depolarization. The rate of fluorescence response of the dye to membrane potential change is usually slower than that of the styryl dye di-4-ANEPPS, but the fluorescence signal change for the former is significantly larger than for the latter dye. DiBAC4(3) has applications in high-throughput drug screening. Properties: Orange solid soluble in DMSO or DMF; Store at -20°C and protect from light; C27H40N4O6; MW: 519; [70363-83-6]
Catalog No. 50-196-4732 Supplier Biotium Supplier No. 61011
Add to Cart
Edge
Add to Cart
DiBAC4(3) (Bis-(1,3-Dibarbituric acid)-trimethine oxanol) is a so-called translational membrane potential dye that redistributes within the cell membrane when membrane potential changes. The fluorescence of the dye is enhanced when the dye enters the cell membrane as a result of membrane depolarization. The rate of fluorescence response of the dye to membrane potential change is usually slower than that of the styryl dye di-4-ANEPPS, but the fluorescence signal change for the former is significantly larger than for the latter dye. DiBAC4(3) has applications in high-throughput drug screening. Properties: Orange solid soluble in DMSO or DMF; Store at -20°C and protect from light; C27H40N4O6; MW: 519; [70363-83-6]