Learn More
Dibasic Sodium Phosphate, Dried, USP, 98-100.5%, Spectrum™ Chemical
Shop All Spectrum Chemical Mfg. Corp. Products

Description
Specifications
Specifications
| CAS Max % | 100% |
| Physical Form | Crystalline Powder or Needles |
| Quantity | 50 kg |
| Percent Purity | 98 to 100.5% |
| Grade | USP |
| Loss on Drying | 5% |
| Packaging | Poly Bottle |
| Chemical Name or Material | Dibasic Sodium Phosphate, Dried |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.