Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dibenamine hydrochloride | 55-43-6 | 99.3% | 1 G
SDP

Supplier:  Medchemexpress LLC HY1283801G

Encompass_Preferred

Dibenamine hydrochloride is a competitive and irreversible adrenergic blocking agent. It is known to modify the pharmacological effects of epinephrine and causes a significant increase in the rate of destruction of I-epinephrine in mice.

  • Competitive and irreversible adrenergic blocking agent
  • Modifies pharmacological effects of epinephrine
  • Increases destruction rate of I-epinephrine in mice
  • Attenuates cocaine-induced increase in sensitivity to acetylcholine
  • Decreases chloroform levels in CCl4-induced hepatotoxicity
  • Slows conversion of CCl4 to chloroform
  • Molecular weight: 296.23
  • Formula: C16H19Cl2N
  • Appearance: White to light yellow solid

Catalog No. 50-003-39119


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.