Learn More
Medchemexpress LLC Dibenamine hydrochloride | 55-43-6 | 99.3% | 5 G
SDP
Catalog No. 5000342620
Encompass_Preferred
Dibenamine hydrochloride is a competitive and irreversible adrenergic blocking agent that modifies the pharmacological effects of epinephrine. It causes a significant increase in the rate of destruction of I-epinephrine in mice.
  • Acts as a competitive and irreversible adrenergic blocking agent.
  • Modifies the pharmacological effects of epinephrine.
  • Increases the rate of destruction of I-epinephrine in mice.
  • Exhibits IC50 for the adrenergic receptor.
  • Attenuates cocaine-induced increases in sensitivity to acetylcholine in vitro.
  • Decreases CHCl3 levels by 30-50% in vivo.
  • Slows the conversion of CCl4 to CHCl3 in vivo.
Catalog No. 50-003-42620 Supplier Medchemexpress LLC Supplier No. HY1283805G
Dibenamine hydrochloride is a competitive and irreversible adrenergic blocking agent that modifies the pharmacological effects of epinephrine. It causes a significant increase in the rate of destruction of I-epinephrine in mice.
  • Acts as a competitive and irreversible adrenergic blocking agent.
  • Modifies the pharmacological effects of epinephrine.
  • Increases the rate of destruction of I-epinephrine in mice.
  • Exhibits IC50 for the adrenergic receptor.
  • Attenuates cocaine-induced increases in sensitivity to acetylcholine in vitro.
  • Decreases CHCl3 levels by 30-50% in vivo.
  • Slows the conversion of CCl4 to CHCl3 in vivo.