Learn More
Medchemexpress LLC Dibenamine hydrochloride (N-(2-Chloroethyl)dibenzylamine hydrochloride) | 55-43-6 | 99.3% | 1 ML
SDP
Catalog No. 5000342618
Encompass_Preferred
Dibenamine hydrochloride is a competitive and irreversible adrenergic blocking agent. It is known for its ability to modify the pharmacological effects of epinephrine and significantly increases the rate of I-epinephrine destruction. In vitro, Dibenamine has been shown to attenuate cocaine-induced increases in sensitivity to acetylcholine in isolated guinea pig vas deferens. In vivo, it has been observed to reduce CHCl3 levels by 30-50 percent, suggesting it slows the conversion of CCl4 to CHCl3.
  • Acts as a competitive and irreversible adrenergic blocking agent
  • Modifies pharmacological effects of epinephrine
  • Increases rate of I-epinephrine destruction
  • Attenuates cocaine-induced increases in sensitivity to acetylcholine in guinea pig vas deferens
  • Reduces CHCl3 levels, slowing CCl4 to CHCl3 conversion
Catalog No. 50-003-42618 Supplier Medchemexpress LLC Supplier No. HY12838010MM/1ML
Dibenamine hydrochloride is a competitive and irreversible adrenergic blocking agent. It is known for its ability to modify the pharmacological effects of epinephrine and significantly increases the rate of I-epinephrine destruction. In vitro, Dibenamine has been shown to attenuate cocaine-induced increases in sensitivity to acetylcholine in isolated guinea pig vas deferens. In vivo, it has been observed to reduce CHCl3 levels by 30-50 percent, suggesting it slows the conversion of CCl4 to CHCl3.
  • Acts as a competitive and irreversible adrenergic blocking agent
  • Modifies pharmacological effects of epinephrine
  • Increases rate of I-epinephrine destruction
  • Attenuates cocaine-induced increases in sensitivity to acetylcholine in guinea pig vas deferens
  • Reduces CHCl3 levels, slowing CCl4 to CHCl3 conversion