Learn More
Selleck Chemical LLC Dibenamine Hydrochloride S5432-25mg
SDP
Catalog No. 503110217
Encompass_Preferred
Dibenamine (N-(2-Chloroethyl)dibenzylamine Sympatholytin Dibenzylchlorethamine) is an irreversible blocker of 1 adrenoceptors
Catalog No. 50-311-0217 Supplier Selleck Chemical LLC Supplier No. S543225MG
Dibenamine (N-(2-Chloroethyl)dibenzylamine Sympatholytin Dibenzylchlorethamine) is an irreversible blocker of 1 adrenoceptors