Learn More
Medchemexpress LLC Dibenz[a,h]anthracene | 53-70-3 | 98.0% | 278.35 | 25 MG
SDP
Catalog No. 5000366396
Encompass_Preferred
Dibenz[a,h]anthracene (DBA) is a polycyclic aromatic hydrocarbon (PAH) with considerable tumorigenicity. It causes DNA adduct formation, leading to the activation of a DNA damage response. DBA induces cell cycle arrest and apoptosis through both Tp53-dependent and Tp53-independent mechanisms.
  • Causes DNA adduct formation
  • Activates a DNA damage response
  • Induces cell cycle arrest
  • Induces apoptosis through both Tp53-dependent and Tp53-independent mechanisms
Catalog No. 50-003-66396 Supplier Medchemexpress LLC Supplier No. 25MGHYW013053
Dibenz[a,h]anthracene (DBA) is a polycyclic aromatic hydrocarbon (PAH) with considerable tumorigenicity. It causes DNA adduct formation, leading to the activation of a DNA damage response. DBA induces cell cycle arrest and apoptosis through both Tp53-dependent and Tp53-independent mechanisms.
  • Causes DNA adduct formation
  • Activates a DNA damage response
  • Induces cell cycle arrest
  • Induces apoptosis through both Tp53-dependent and Tp53-independent mechanisms