Learn More
Medchemexpress LLC Dibenz[a,h]anthracene | 53-70-3 | 98.0% | 278.35 | 50 MG
SDP
Catalog No. 5000451235
Encompass_Preferred
Dibenz[a,h]anthracene (DBA) is a polycyclic aromatic hydrocarbon (PAH) known for its tumorigenicity. It induces DNA adduct formation, triggering a DNA damage response. The compound also leads to cell cycle arrest and apoptosis via both Tp53-dependent and Tp53-independent pathways, making it suitable for research applications.
  • Considerable tumorigenicity
  • Causes DNA adduct formation
  • Activates a DNA damage response
  • Induces cell cycle arrest
  • Induces apoptosis through Tp53-dependent and Tp53-independent mechanisms
  • For research use only
Catalog No. 50-004-51235 Supplier Medchemexpress LLC Supplier No. HYW01305350MG
Dibenz[a,h]anthracene (DBA) is a polycyclic aromatic hydrocarbon (PAH) known for its tumorigenicity. It induces DNA adduct formation, triggering a DNA damage response. The compound also leads to cell cycle arrest and apoptosis via both Tp53-dependent and Tp53-independent pathways, making it suitable for research applications.
  • Considerable tumorigenicity
  • Causes DNA adduct formation
  • Activates a DNA damage response
  • Induces cell cycle arrest
  • Induces apoptosis through Tp53-dependent and Tp53-independent mechanisms
  • For research use only