Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibenz[a,h]anthracene, 100 μg/mL, Methylene Chloride, 1 mL

Catalog No. USP7001
Encompass
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-P7001 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-P7001 Supplier Agilent Technologies Supplier No. P7001
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 53-70-3 , 75-09-2
Molecular Formula C22H14
Molecular Weight (g/mol) 278.354
InChI Key LHRCREOYAASXPZ-UHFFFAOYSA-N
PubChem CID 5889
ChEBI CHEBI:35299
IUPAC Name naphtho[1,2-b]phenanthrene
SMILES C1=CC=C2C(=C1)C=CC3=CC4=C(C=CC5=CC=CC=C54)C=C32

Specifications

CAS Max % 99.99, 0.01
Physical Form Liquid
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 100μg/mL
Chemical Name or Material Dibenz[a, h]anthracene Solution, 100μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.