Learn More
Selleck Chemical LLC Dibenzazepine (YO-01027) 2mg 209984-56-5
SDP
Supplier:  Selleck Chemical LLC S27112MG
Encompass_Preferred
Dibenzazepine (YO-01027) is a dipeptidic ?-secretase inhibitor with IC50 of 2.6 nM and 2.9 nM in cell-free assays for APPL and Notch cleavage, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-2816