Learn More
Selleck Chemical LLC Diphenyleneiodonium chloride (DPI) 10mg 4673-26-1
SDP
Catalog No. 501948039
Encompass_Preferred
Diphenyleneiodonium chloride (DPI) is an inhibitor of NADPH oxidase and also a potent, irreversible, and time-, temperature-dependent iNOS/eNOS inhibitor. Diphenyleneiodonium chloride (DPI) also functions as a TRPA1 activator and selectively inhibits intracellular reactive oxygen species (ROS). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8039 Supplier Selleck Chemical LLC Supplier No. S863910MG
Diphenyleneiodonium chloride (DPI) is an inhibitor of NADPH oxidase and also a potent, irreversible, and time-, temperature-dependent iNOS/eNOS inhibitor. Diphenyleneiodonium chloride (DPI) also functions as a TRPA1 activator and selectively inhibits intracellular reactive oxygen species (ROS). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.