Learn More
Strem, An Ascensus Company CAS# 14187-32-7. 10g. Dibenzo-18-crown-6, min. 98%. MFCD00005098
SDP
Catalog No. 5090109174
Encompass
CAS# 14187-32-7. 10g. Dibenzo-18-crown-6, min. 98%. MFCD00005098. Molecular Weight: 360.41. Molecular Formula: C20H24O6. Color/form: white pwdr. Strem# 08-0500. http://www.strem.com/catalog/v/08-0500/
Catalog No. 50-901-09174 Supplier Strem, An Ascensus Company Supplier No. 08050010G
CAS# 14187-32-7. 10g. Dibenzo-18-crown-6, min. 98%. MFCD00005098. Molecular Weight: 360.41. Molecular Formula: C20H24O6. Color/form: white pwdr. Strem# 08-0500. http://www.strem.com/catalog/v/08-0500/