Learn More
Medchemexpress LLC Dibenzo-24-crown-8-e | 1g
SDP
Supplier:  Medchemexpress LLC HYW0731831G
Encompass_Preferred
Dibenzo-24-crown-8-ether is a phase transfer catalyst that can reduce H2PtCl6 6H2O and FeCl2 4H2O in a thermal system to synthesize 17 nm monodispersed iron-platinum (FePt) alloy nanoparticles[1]
Catalog No. 50-002-10488