Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dibenzo[a,g]quinolizinium, 5,6-dihydro-2-hydroxy-3,9,10-trimethoxy-, chloride (1:1) | 1916-10-5 | 99.6% | 10 MG
SDP

Catalog No. 5000296536
Encompass_Preferred

Columbamine chloride is the chloride salt of columbamine, a benzylisoquinoline alkaloid used in biological research. Supplied as a light-yellow solid with high reported purity, it is intended for in vitro screening and preclinical studies; follow institutional safety procedures for handling and storage.

  • Molecular formula C20H20ClNO4.
  • Molecular weight 373.83.
  • Appearance light-yellow solid.
  • Purity 99.62% (LCMS).
  • Storage 4°C, sealed, protect from moisture and light.
  • Typical applications include in vitro antioxidant, anti-inflammatory, and antitumor screening.

Catalog No. 50-002-96536 Supplier Medchemexpress LLC Supplier No. HYN0926A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Columbamine chloride is the chloride salt of columbamine, a benzylisoquinoline alkaloid used in biological research. Supplied as a light-yellow solid with high reported purity, it is intended for in vitro screening and preclinical studies; follow institutional safety procedures for handling and storage.

  • Molecular formula C20H20ClNO4.
  • Molecular weight 373.83.
  • Appearance light-yellow solid.
  • Purity 99.62% (LCMS).
  • Storage 4°C, sealed, protect from moisture and light.
  • Typical applications include in vitro antioxidant, anti-inflammatory, and antitumor screening.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.