Learn More
Medchemexpress LLC Dibenzo[b,d]furan | 132-64-9 | MFCD00004968 | 100.0% | 168.19 g/mol | C12H8O | 25 G
SDP
Catalog No. 5000303070
Encompass_Preferred
Dibenzo[b,d]furan is an aromatic heterocyclic organic compound used as a biochemical reagent in research and analytical workflows. It serves as a stable polycyclic heteroarene for organic synthesis, method development, and analytical standardization. The compound has the formula C12H8O and a molecular weight of 168.19 g/mol.
  • Chemical formula C12H8O and molecular weight 168.19 g/mol.
  • High purity suitable for research and analytical use.
  • Stable at room temperature; extended storage life in solvent at -80 °C or -20 °C.
  • Applicable for organic synthesis, analytical standards, and biochemical studies.
  • Supplied with datasheet, certificate of analysis, and safety data sheet for quality and handling information.
Catalog No. 50-003-03070 Supplier Medchemexpress LLC Supplier No. 25GHYW017086
Dibenzo[b,d]furan is an aromatic heterocyclic organic compound used as a biochemical reagent in research and analytical workflows. It serves as a stable polycyclic heteroarene for organic synthesis, method development, and analytical standardization. The compound has the formula C12H8O and a molecular weight of 168.19 g/mol.
  • Chemical formula C12H8O and molecular weight 168.19 g/mol.
  • High purity suitable for research and analytical use.
  • Stable at room temperature; extended storage life in solvent at -80 °C or -20 °C.
  • Applicable for organic synthesis, analytical standards, and biochemical studies.
  • Supplied with datasheet, certificate of analysis, and safety data sheet for quality and handling information.