Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dibenzo[b,d]furan | 132-64-9 | MFCD00004968 | 100.0% | 168.19 g·mol⁻¹ | C12H8O | 50 G
SDP

Catalog No. 5000301483
Encompass_Preferred

Dibenzo[b,d]furan (CAS 132-64-9) is an aromatic heterocyclic compound used as a biochemical reagent and organic building block in life-science research. Supplied as a solid at high purity, it is intended for analytical and synthetic laboratory applications.

  • High purity (99.95%).
  • Molecular formula C12H8O.
  • Molecular weight 168.19 g·mol⁻¹.
  • Supplied as a solid suitable for handling and storage.
  • Available in multiple pack sizes for laboratory use.
  • Suitable for analytical and synthetic applications in life-science research.

Catalog No. 50-003-01483 Supplier Medchemexpress LLC Supplier No. HYW01708650G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dibenzo[b,d]furan (CAS 132-64-9) is an aromatic heterocyclic compound used as a biochemical reagent and organic building block in life-science research. Supplied as a solid at high purity, it is intended for analytical and synthetic laboratory applications.

  • High purity (99.95%).
  • Molecular formula C12H8O.
  • Molecular weight 168.19 g·mol⁻¹.
  • Supplied as a solid suitable for handling and storage.
  • Available in multiple pack sizes for laboratory use.
  • Suitable for analytical and synthetic applications in life-science research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.