Learn More
Medchemexpress LLC Dibenzo[b,d]furan-d8 | 93952-04-6 | 99.9% | 176.24 g/mol | C12D8O | 100 MG
SDP
Catalog No. 5000301819
Encompass_Preferred
Dibenzo[b,d]furan-d8 is the deuterium-labeled isotopologue of dibenzofuran supplied as a high-purity stable-isotope reference for analytical and research applications. It is intended for use as a tracer or internal standard in mass spectrometry and environmental contaminant testing, provided as a solid with specified isotopic labeling and purity, for research use only.
  • High isotopic enrichment and purity.
  • Suitable as a tracer or internal standard for mass spectrometry.
  • Stable solid form for storage and handling.
  • Available in small pack sizes compatible with analytical workflows.
  • For research use only; not for human or clinical use.
Catalog No. 50-003-01819 Supplier Medchemexpress LLC Supplier No. HYW017086S100MG
Dibenzo[b,d]furan-d8 is the deuterium-labeled isotopologue of dibenzofuran supplied as a high-purity stable-isotope reference for analytical and research applications. It is intended for use as a tracer or internal standard in mass spectrometry and environmental contaminant testing, provided as a solid with specified isotopic labeling and purity, for research use only.
  • High isotopic enrichment and purity.
  • Suitable as a tracer or internal standard for mass spectrometry.
  • Stable solid form for storage and handling.
  • Available in small pack sizes compatible with analytical workflows.
  • For research use only; not for human or clinical use.