Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibenzo[b,d]thiophen-2-ylboronic acid, 97%, Thermo Scientific™

Catalog No. MO01033CB
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MO01033CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MO01033CB Supplier Thermo Scientific Chemicals Supplier No. MO01033CB
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 668983-97-9
Molecular Formula C12H9BO2S
Molecular Weight (g/mol) 228.072
MDL Number MFCD01318982
InChI Key CSLSCVHILGCSTE-UHFFFAOYSA-N
Synonym dibenzo b,d thiophen-2-ylboronic acid, dibenzothiophene-2-boronic acid, dibenzo b,d thien-2-ylboronic acid, dibenzothiophen-2-yl-dihydroxy-borane, boronic acid, 2-dibenzothienyl, boronic acid, b-2-dibenzothienyl, dibenzo b,d thiophen-2-ylboronic acid contains varying amounts of anhydride, dibenzo b,d thien-2-ylboronic acid saltdata: free, 8-thiatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaen-4-ylboronic acid, ksc352o5l
PubChem CID 2794660
IUPAC Name dibenzothiophen-2-ylboronic acid
SMILES B(C1=CC2=C(C=C1)SC3=CC=CC=C32)(O)O

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 228.08
Percent Purity 95%
Chemical Name or Material Dibenzo[b, d]thiophen-2-ylboronic acid, May contain varying amounts of anhydride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.