Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dibenzo[b,e]oxepin-11(6H)-one | 4504-87-4 | 210.23 | 10 G
SDP

Catalog No. 5000446204
Encompass_Preferred

Dibenzo[b,e]oxepin-11(6H)-one is a dibenzoxepinone derivative demonstrating significant antiparasitic activity. This compound is intended for research use only and exhibits inhibitory effects on the motility of the nematode *Caenorhabditis elegans*. A concentration of 1 mM can reduce worm thrashes by 80% within 10 minutes.

  • Antiparasitic activity
  • Inhibits *Caenorhabditis elegans* motility
  • For research use only
  • Appearance: Solid
  • Color: Off-white to light yellow
  • Storage: Room temperature for 3 years; in solvent, -80°C for 2 years, -20°C for 1 year

Catalog No. 50-004-46204 Supplier Medchemexpress LLC Supplier No. HYW00667210G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dibenzo[b,e]oxepin-11(6H)-one is a dibenzoxepinone derivative demonstrating significant antiparasitic activity. This compound is intended for research use only and exhibits inhibitory effects on the motility of the nematode *Caenorhabditis elegans*. A concentration of 1 mM can reduce worm thrashes by 80% within 10 minutes.

  • Antiparasitic activity
  • Inhibits *Caenorhabditis elegans* motility
  • For research use only
  • Appearance: Solid
  • Color: Off-white to light yellow
  • Storage: Room temperature for 3 years; in solvent, -80°C for 2 years, -20°C for 1 year

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.