Learn More
Medchemexpress LLC Dibenzo[b,e]oxepin-11(6H)-one | 4504-87-4 | 210.23 | 25 G
SDP
Catalog No. 5000448221
Encompass_Preferred
Dibenzo[b,e]oxepin-11(6H)-one is a dibenzoxepinone derivative known for its significant antiparasitic activity. This compound is intended for research use only.
  • Inhibits motility of the nematode *Caenorhabditis elegans*
  • Reduces worm's thrashes by 80% at 1 mM concentration for 10 minutes
Catalog No. 50-004-48221 Supplier Medchemexpress LLC Supplier No. 25GHYW006672
Dibenzo[b,e]oxepin-11(6H)-one is a dibenzoxepinone derivative known for its significant antiparasitic activity. This compound is intended for research use only.
  • Inhibits motility of the nematode *Caenorhabditis elegans*
  • Reduces worm's thrashes by 80% at 1 mM concentration for 10 minutes