Learn More
Medchemexpress LLC Dibenzo[b,e]oxepin-11(6H)-one | 4504-87-4 | 210.23 | 5 G
SDP
Catalog No. 5000446558
Encompass_Preferred
Dibenzo[b,e]oxepin-11(6H)-one is a dibenzoxepinone derivative known for its significant antiparasitic activity. It is intended for research use only.
  • Exhibits inhibitory effects on the motility of the nematode *Caenorhabditis elegans*
  • Treatment with 1 mM can reduce worm thrashing by 80%
  • Off-white to light yellow solid appearance
Catalog No. 50-004-46558 Supplier Medchemexpress LLC Supplier No. HYW0066725G
Dibenzo[b,e]oxepin-11(6H)-one is a dibenzoxepinone derivative known for its significant antiparasitic activity. It is intended for research use only.
  • Exhibits inhibitory effects on the motility of the nematode *Caenorhabditis elegans*
  • Treatment with 1 mM can reduce worm thrashing by 80%
  • Off-white to light yellow solid appearance