Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibenzo[b,f][1,4]thiazepin-11(10H)-one 98.0+%, TCI America™
SDP

Catalog No. D369325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D369325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D369325G Supplier TCI America Supplier No. D369325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7-7-3159
Molecular Formula C13H9NOS
Molecular Weight (g/mol) 227.28
MDL Number MFCD00901197
InChI Key RTERDTBXBYNZIS-UHFFFAOYSA-N
PubChem CID 374829
IUPAC Name 5H-benzo[b][1,4]benzothiazepin-6-one
SMILES C1=CC=C2C(=C1)C(=O)NC3=CC=CC=C3S2

Specifications

Melting Point 265°C
Color White-Yellow
Quantity 25 g
Formula Weight 227.28
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Dibenzo[b,f][1,4]thiazepin-11(10H)-one
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.