Learn More
Medchemexpress LLC Dibenzo[b,k]hexaoxacyclooctadecin, eicosahydro- | 16069-36-6 | 97.0% | 372.50 | 250 MG
SDP
Catalog No. 5000451374
Encompass_Preferred
Dicyclohexano-18-crown-6-ether selectively extracts lysine-rich cytochrome c proteins from other cationic proteins under weakly acidic and neutral conditions, demonstrating specificity in ligand-protein interactions.
  • Selectively extracts lysine-rich cytochrome c proteins from other cationic proteins.
  • Demonstrates specificity in ligand-protein interactions.
  • Shows antiproliferative activity against various cell lines (HeLa, MCF7, MIA PaCa-2, NCI-H460, SW-620).
Catalog No. 50-004-51374 Supplier Medchemexpress LLC Supplier No. 250MGHYW009048
Dicyclohexano-18-crown-6-ether selectively extracts lysine-rich cytochrome c proteins from other cationic proteins under weakly acidic and neutral conditions, demonstrating specificity in ligand-protein interactions.
  • Selectively extracts lysine-rich cytochrome c proteins from other cationic proteins.
  • Demonstrates specificity in ligand-protein interactions.
  • Shows antiproliferative activity against various cell lines (HeLa, MCF7, MIA PaCa-2, NCI-H460, SW-620).