Learn More
Medchemexpress LLC Dibenzocyclooctyne-amine | 1255942-06-3 | 276.33 | 50 MG
SDP
Catalog No. 5000447108
Encompass_Preferred
DBCO-amine (GMP) is a GMP grade DBCO-amine (HY-W000423). DBCO-amine is a click chemistry reagent, containing a DBCO group that can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing Azide groups. It is used in the synthesis of antibody-drug conjugates (ADCs).
Catalog No. 50-004-47108 Supplier Medchemexpress LLC Supplier No. HYW000423G50MG
DBCO-amine (GMP) is a GMP grade DBCO-amine (HY-W000423). DBCO-amine is a click chemistry reagent, containing a DBCO group that can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing Azide groups. It is used in the synthesis of antibody-drug conjugates (ADCs).