Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dibenzocyclooctyne-PEG-thiol | 95.0% | 3400.00 | 10 MG
SDP

Catalog No. 5000297689
Encompass_Preferred

DBCO-PEG-SH is a dibenzocyclooctyne-functionalized polyethylene glycol bearing a terminal thiol. It is used as a bifunctional linker for strain-promoted alkyne-azide cycloaddition (SPAAC) and thiol-based coupling in biological and materials research. Supplied as a solid, it has an approximate molecular weight of 3400 Da and 95.0% purity.

  • Dibenzocyclooctyne-functionalized PEG with terminal thiol for bioconjugation.
  • Molecular weight ~3400 Da and 95.0% purity.
  • Soluble in DMSO at 100 mg/mL; requires ultrasonic agitation.
  • Solid, off-white to light yellow appearance.
  • Store at 4°C and protect from light; in solution, store at -80°C up to 6 months.

Catalog No. 50-002-97689 Supplier Medchemexpress LLC Supplier No. HYD230610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DBCO-PEG-SH is a dibenzocyclooctyne-functionalized polyethylene glycol bearing a terminal thiol. It is used as a bifunctional linker for strain-promoted alkyne-azide cycloaddition (SPAAC) and thiol-based coupling in biological and materials research. Supplied as a solid, it has an approximate molecular weight of 3400 Da and 95.0% purity.

  • Dibenzocyclooctyne-functionalized PEG with terminal thiol for bioconjugation.
  • Molecular weight ~3400 Da and 95.0% purity.
  • Soluble in DMSO at 100 mg/mL; requires ultrasonic agitation.
  • Solid, off-white to light yellow appearance.
  • Store at 4°C and protect from light; in solution, store at -80°C up to 6 months.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.