Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dibenzocyclooctyne-Peg4-Biotin Conjugate

Catalog No. 7607495MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-074-95MG 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 76-074-95MG Supplier Sigma Aldrich Supplier No. 7607495MG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

This azadibenzocyclooctyne-biotin derivative is a versatile biotinylation reagent for labeling azide containing molecules or compounds. Cyclooctynes are useful in strain-promoted copper-free azide-alkyne cycloaddition reactions.This dibenzocyclooctyne will react with azide-functionalized compounds or biomolecules without the need for a Cu(I) catalyst to result in a stable triazole linkage. Subsequent labeled molecules can be purified using streptavidin or avidin affinity reagents. The PEG spacer makes the overall compound more hydrophilic and allows for greater distance between the reaction site with the azide and the biotin detecting group.activity-based protein profilingdetection of newly synthesized proteinsimmobilization of azide-labeled biomolecules

Specifications

CAS 26391-06-0
Density 1.013 g/mL at 25°C
Molecular Formula C7H12N2O
Linear Formula C7H12N2O
MDL Number MFCD01332383
Quantity 5 g
Molecular Weight (g/mol) 140.18 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.